Anti-PIP4K2C Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP8043AEA 405 CHF
ABGEAP8043A
Anti-PIP4K2C Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: PIP4K2C (phosphatidylinositol-5-phosphate 4-kinase, type II, gamma)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR