Anti-PIP4K2A Rabbit Polyclonal Antibody

Supplier: Abgent
ABGEAP8041AEA 378 CHF
ABGEAP8041A
Anti-PIP4K2A Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: PIP4K2A (phosphatidylinositol-5-phosphate 4-kinase, type II, alpha)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR