Anti-PIP5K1C Rabbit Polyclonal Antibody

Supplier: Abgent
ABGEAP8039AEA 419 CHF
ABGEAP8039A
Anti-PIP5K1C Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: PIP5K1C (phosphatidylinositol-4-phosphate 5-kinase, type I, gamma)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR