Anti-PIP5K1A Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP8037AEA 405 CHF
ABGEAP8037A
Anti-PIP5K1A Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: PIP5K1A (phosphatidylinositol-4-phosphate 5-kinase, type I, alpha)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR