Anti-GNA12 Rabbit Polyclonal Antibody

Supplier: Abgent
ABGEAP8035AEA 378 CHF
ABGEAP8035A
Anti-GNA12 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: GNA12 (guanine nucleotide binding protein (G protein) alpha 12)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR