Anti-Endophilin B1 Rabbit Polyclonal Antibody
Lieferant: Abgent
ABGEAP8034AEA 405 CHF
ABGEAP8034A
Anti-Endophilin B1 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: Endophilin B1
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human