Anti-PIK3R2 Rabbit Polyclonal Antibody

Supplier: Abgent
ABGEAP8028AEA 407 CHF
ABGEAP8028A
Anti-PIK3R2 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: PIK3R2 (phosphoinositide-3-kinase, regulatory subunit 2 (beta))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR