Anti-PI3 Kinase p85 (PIK3R3) Rabbit Polyclonal Antibody

Supplier: Abgent
ABGEAP8025AEA 431 CHF
ABGEAP8025A
Anti-PI3 Kinase p85 (PIK3R3) Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: PI3 Kinase p85 (PIK3R3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR