Anti-PIK3R1 Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP8023FEA 405 CHF
ABGEAP8023F
Anti-PIK3R1 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: PIK3R1 (phosphoinositide-3-kinase, regulatory subunit 1 (alpha))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR