Anti-PI3KCD Rabbit Polyclonal Antibody

Supplier: Abgent
ABGEAP8020DEA 407 CHF
ABGEAP8020D
Anti-PI3KCD Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: PI3KCD
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR