Anti-PI3KC2G Rabbit Polyclonal Antibody

Supplier: Abgent
ABGEAP8012AEA 407 CHF
ABGEAP8012A
Anti-PI3KC2G Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: PI3KC2G
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR