Anti-RPS6KB2 Rabbit Polyclonal Antibody

Supplier: Abgent
ABGEAP8009EEA 448 CHF
ABGEAP8009E
Anti-RPS6KB2 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: RPS6KB2 (ribosomal protein S6 kinase, 70kDa, polypeptide 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR