Anti-MAP3K13 (LZK) Rabbit Polyclonal Antibody

Supplier: Abgent
ABGEAP8008DEA 407 CHF
ABGEAP8008D
Anti-MAP3K13 (LZK) Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: MAP3K13 (LZK)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR