Anti-MAP3K14 Rabbit Polyclonal Antibody

Supplier: Abgent
ABGEAP8004AEA 419 CHF
ABGEAP8004A
Anti-MAP3K14 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: MAP3K14 (mitogen-activated protein kinase kinase kinase 14)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR