Anti-CYP7A1 Rabbit Polyclonal Antibody

Supplier: Abgent
ABGEAP7996AEA 378 CHF
ABGEAP7996A
Anti-CYP7A1 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: CYP7A1 (cytochrome P450, family 7, subfamily A, polypeptide 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR