Anti-CYP1A1 Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP7993CEA 405 CHF
ABGEAP7993C
Anti-CYP1A1 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: CYP1A1 (cytochrome P450, family 1, subfamily A, polypeptide 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR