Anti-CYP1A1 Rabbit Polyclonal Antibody

Supplier: Abgent
ABGEAP7993BEA 448 CHF
ABGEAP7993B
Anti-CYP1A1 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: CYP1A1 (cytochrome P450, family 1, subfamily A, polypeptide 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR