Anti-TIPARP Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP7978AEA 405 CHF
ABGEAP7978A
Anti-TIPARP Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: TIPARP (TCDD-inducible poly(ADP-ribose) polymerase)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR