Solvent bottles, glass coated
Lieferant: Thermo Fisher Scientific
Dionex™
DION045901EA 0 CHF
DION045901
Solvent bottles, glass coated
Flaschen HPLC-Flaschen
Borosilicate glass, coated.
The solvent bottle and cap assembly holds organic solvents or aqueous buffers to be used as extraction solvents for ASE® systems. The quick-connect fittings allow for easy assembly of the bottles to the systems.