U-tube capillary viscometers, BS/IP/RF
Lieferant: Fungilab
FUNGCV006-010EA 207 CHF
FUNGCV006-010 FUNGCV006-011 FUNGCV006-001 FUNGCV006-006 FUNGCV006-007 FUNGCV006-008 FUNGCV006-009 FUNGCV006-002 FUNGCV006-003 FUNGCV006-004 FUNGCV006-005
U-tube capillary viscometers, BS/IP/RF
Viskosimeter Kapillarviskosimeter
Capillary viscometers with indelible timing marks and reverse flow for opaque liquids.
  • Constant value at 40 ºC and 100 ºC into the calibration certificate
  • Precision: ±0,3 %
  • Borosilicate glass
Liquid is drawn into the upper bulb by suction, then flowed down through the capillary into the lower bulb.
Zertifizierungen: ASTM D-445, D446, ISO 3104, 3105 and UNE 400313
Lieferumfang: Supplied with the calibration certificate issued in accordance with the conditions of accreditation granted by a National Metrological Register, the test uncertainty and its traceability to international standards UKAS.