Sterilisation boxes for pipettes, aluminium
Lieferant: Nickel-Electro
NICK6002EA 138 CHF
NICK6002 NICK6001
Sterilisation boxes for pipettes, aluminium
Round sterilising pipette cans made of aluminium.
  • Excellent chemical resistance
  • Lightweight
  • Sturdy
They can be steam sterilised at 120 °C and withstand dry heat temperatures of up to 180 °C.