Refrigerated re-circulation thermostat, Thermovisc 100-F8
Lieferant: Fungilab
FUNGB100F8EA 5930 CHF
FUNGB100F8
Refrigerated re-circulation thermostat, Thermovisc 100-F8
Refrigerated re-circulating thermostat made of AISI 304 stainless steel top and lid with an inner tank of AISI 310 stainless steel.
Complete with a pump for internal and external liquid circulation with push on tube connectors for inlet and outlet. A drain tap is placed at the side of the unit. A hermetically sealed CFC free compressor with a ventilated evaporator is mounted on anti-vibration mounts.