2-Hydroxy-3-nitro-4-picolin ≥98%

Achtung

Synonyms: 4-Methyl-3-nitro-2-pyridon2-Hydroxy-4-methyl-3-nitropyridin

H61586.06EA 222 CHF
H61586.06
2-Hydroxy-3-nitro-4-picolin ≥98%
2-Hydroxy-3-nitro-4-picolin
Formel: C₆H₆N₂O₃
MW: 154,13 g/mol
Melting Pt: 229…232 °C
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00010689
CAS-Nummer: 21901-18-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR