1-(Phenylsulfonyl)-6-indolboronsäure
Lieferant: ACTIVATE SCIENTIFIC GMBH
ACTIAS7234G5EA
0
CHF
ACTIAS7234G5
ACTIAS7234G25
ACTIAS7234G1
1-(Phenylsulfonyl)-6-indolboronsäure
1-(Phenylsulphonyl)-6-indoleboronic acid
Formel:
C₁₄H₁₂BNO₄S MW: 301,1254 g/mol |
MDL Number:
MFCD03840611 CAS-Nummer: 480438-52-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...