DNA extraction kits, ZyGEM™ prepGEM® Storage Card Blood kit
ZYGEPSB0050EA 204 CHF
ZYGEPSB0050 ZYGEPSB0200
DNA extraction kits, ZyGEM™ prepGEM® Storage Card Blood kit
prepGEM® DNA extraction kits have been developed to provide a rapid, reliable, and economical solution to DNA extraction. The kits use a single tube protocol with temperature control to prepare DNA for analysis, and are optimised for downstream PCR-based applications, including qPCR, STR-PCR, DNA sequencing, AFLP analysis, and many SNiP detection applications.
prepGEM® Storage Card Blood kit has been specifically developed for extracting DNA from human or mouse blood stored on FTA cards and is particularly valuable for high throughput automation of high volume crime samples. The procedure is easily adaptable for 96-well plates and automation, and the closed-tube nature of the method protects the integrity of the sample through the extraction procedure.
prepGEM® Storage Card Blood kit has been specifically developed for extracting DNA from human or mouse blood stored on FTA cards and is particularly valuable for high throughput automation of high volume crime samples. The procedure is easily adaptable for 96-well plates and automation, and the closed-tube nature of the method protects the integrity of the sample through the extraction procedure.