Infrared hotplate, SLK 12
Lieferant: SI Analytics
453-0141EA 840 CHF
SCOI285416630
Infrared hotplate, SLK 12
Infrared hotplate with unpainted stainless steel casing and ceramic glass heating surface. Unit is easy to clean, resistant to chemical influences and temperature fluctuations. The ceramic glass heating surface has a high infrared permeability and so is exceptionally economic, saving both energy and time. The heating power can be adjusted in 9 steps and heats up to 1,7 kW (with extended hotplate step 2). The heating surface is planar throughout and offers unheated border areas. This allows the vessel to be heated partially or completely removed from the heating area.
  • Heating area with two zones for small and larger vessels
  • LED indicator shows if there is residual heat to ensure safe operation
  • Optional compressed air connection allows operation in highly aggressive environments