4-Chlor-4'-hydroxybenzophenon

Lieferant: ACTIVATE SCIENTIFIC GMBH

ACTIAS22002G100EA 0 CHF
ACTIAS22002G100
4-Chlor-4'-hydroxybenzophenon
4-Chlor-4'-hydroxybenzophenon
Formel: C₁₃H₉ClO₂
MW: 232,67 g/mol
MDL Number: MFCD00002357
CAS-Nummer: 42019-78-3
EINECS: 255-627-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR