5-Amino-3-(3-bromphenyl)-1H-pyrazol
Lieferant: ACTIVATE SCIENTIFIC GMBH
ACTIAS23819G25EA
0
CHF
ACTIAS23819G25
ACTIAS23819G5
5-Amino-3-(3-bromphenyl)-1H-pyrazol
5-Amino-3-(3-bromphenyl)-1H-pyrazol
Formel:
C₉H₈BrN₃ |
MDL Number:
MFCD06738756 CAS-Nummer: 149246-81-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...