4-Iod-1-methyl-1H-pyrazol

Lieferant: ACTIVATE SCIENTIFIC GMBH

ACTIAS12910G100EA 119 CHF
ACTIAS12910G100
4-Iod-1-methyl-1H-pyrazol
4-Iodo-1-methyl-1H-pyrazole
Formel: C₄H₅IN₂
MW: 208,0000 g/mol
MDL Number: MFCD05663860
CAS-Nummer: 39806-90-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR