3,4-(Methylenedioxy)phenylboronsäure
Lieferant: ACTIVATE SCIENTIFIC GMBH
ACTIAS50876G5EA
0
CHF
ACTIAS50876G5
ACTIAS50876G25
ACTIAS50876G1
3,4-(Methylenedioxy)phenylboronsäure
3,4-(Methylenedioxy)phenylboronsäure
Formel:
C₇H₇BO₄ MW: 165,94 g/mol |
MDL Number:
MFCD01009695 CAS-Nummer: 94839-07-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...