4-Amino-1-Boc-piperidin-4-carbonsäure

Lieferant: ACTIVATE SCIENTIFIC GMBH

ACTIAS21089G5EA 98 CHF
ACTIAS21089G5 ACTIAS21089G1
4-Amino-1-Boc-piperidin-4-carbonsäure
4-Amino-1-Boc-piperidin-4-carbonsäure
Formel: C₁₁H₂₀N₂O₄
MW: 244,29 g/mol
MDL Number: MFCD01318728
CAS-Nummer: 183673-71-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR