Ethylacetopyruvat

Lieferant: ACTIVATE SCIENTIFIC GMBH

ACTIAS37549G5EA 0 CHF
ACTIAS37549G10 ACTIAS37549G5
Ethylacetopyruvat
Ethylacetopyruvat
Formel: C₇H₁₀O₄
MW: 158,15 g/mol
MDL Number: MFCD00009124
CAS-Nummer: 615-79-2
EINECS: 210-447-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR