2-Methylnicotinamid

Lieferant: ACTIVATE SCIENTIFIC GMBH

ACTIAS32608G1EA 0 CHF
ACTIAS32608G1 ACTIAS32608G5
2-Methylnicotinamid
2-Methylnicotinamid
Formel: C₇H₈N₂O
MW: 136,1512 g/mol
MDL Number: MFCD09909457
CAS-Nummer: 58539-65-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR