3-Formyl-4-methoxyphenylboronsäure
Lieferant: ACTIVATE SCIENTIFIC GMBH
ACTIAS51441G100EA
0
CHF
ACTIAS51441G100
ACTIAS51441G5
ACTIAS51441G1
ACTIAS51441G25
3-Formyl-4-methoxyphenylboronsäure
3-Formyl-4-methoxyphenylboronsäure
Formel:
C₈H₉BO₄ MW: 179,97 g/mol |
MDL Number:
MFCD02093661 CAS-Nummer: 121124-97-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...