3-(Hydroxymethyl)phenylboronsäurepinakolester
Lieferant: ACTIVATE SCIENTIFIC GMBH
Synonyms:
3-(Hydroxymethyl)benzolboronsäure(tetramethylethylen)ester
ACTIAS51367G1EA
0
CHF
ACTIAS51367G25
ACTIAS51367G1
ACTIAS51367G5
3-(Hydroxymethyl)phenylboronsäurepinakolester
3-(Hydroxymethyl)phenylboronsäurepinakolester
Formel:
C₁₃H₁₉BO₃ MW: 234,10 g/mol |
MDL Number:
MFCD09266196 CAS-Nummer: 443776-76-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...