2,6-Dimethylbenzaldehyde

Supplier: ACTIVATE SCIENTIFIC GMBH

ACTIAS1477G5EA 0 CHF
ACTIAS1477G25 ACTIAS1477G5
2,6-Dimethylbenzaldehyde
2,6-Dimethylbenzaldehyde
Formula: C₉H₁₀O
MW: 134.18 g/mol
MDL Number: MFCD00128003
CAS Number: 1123-56-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR