Isobutyramide

Supplier: ACTIVATE SCIENTIFIC GMBH

ACTIAS50924G100EA 0 CHF
ACTIAS50924G25 ACTIAS50924G100
Isobutyramide
Isobutyramide
Formula: C₄H₉NO
MW: 87.12 g/mol
MDL Number: MFCD00008019
CAS Number: 563-83-7
EINECS: 209-265-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR