2-Methoxy-5-pyridineboronic acid

Supplier: ACTIVATE SCIENTIFIC GMBH

Synonyms: (6-Methoxypyridin-3-yl)boronic acid

ACTIAS50874G5EA 0 CHF
ACTIAS50874G25 ACTIAS50874G1 ACTIAS50874G5
2-Methoxy-5-pyridineboronic acid
2-Methoxy-5-pyridineboronic acid
Formula: C₆H₈BNO₃
MW: 152.95 g/mol
MDL Number: MFCD02093044
CAS Number: 163105-89-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR