5-Fluor-3-methylbenzo[b]thiophen
Lieferant: ACTIVATE SCIENTIFIC GMBH
ACTIAS35284G1EA
0
CHF
ACTIAS35284G1
ACTIAS35284G10
ACTIAS35284G5
5-Fluor-3-methylbenzo[b]thiophen
5-Fluor-3-methylbenzo[b]thiophen
Formel:
C₉H₇FS |
MDL Number:
MFCD00179348 CAS-Nummer: 17514-63-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...