5-Aminophthalazine

Supplier: ACTIVATE SCIENTIFIC GMBH

ACTIAS30342G5EA 0 CHF
ACTIAS30342G5 ACTIAS30342G1
5-Aminophthalazine
5-Aminophthalazine
Formula: C₈H₇N₃
MW: 145.1613 g/mol
MDL Number: MFCD08272072
CAS Number: 102072-84-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR