4-Bromo-2-(trifluoromethyl)quinoline

ACTIAS2542G1EA 0 CHF
ACTIAS2542G1
4-Bromo-2-(trifluoromethyl)quinoline
4-Bromo-2-(trifluoromethyl)quinoline
Formula: C₁₀H₅BrF₃N
MW: 276.0526 g/mol
MDL Number: MFCD00277488
CAS Number: 18706-25-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR