Benzo[b]furan-2-boronic acid

Supplier: ACTIVATE SCIENTIFIC GMBH

Synonyms: Benzofuran-2-boronic acid

ACTIAS50225G5EA 0 CHF
ACTIAS50225G5 ACTIAS50225G100 ACTIAS50225G25
Benzo[b]furan-2-boronic acid
Benzo[b]furan-2-boronic acid
Formula: C₈H₇BO₃
MW: 161.95 g/mol
MDL Number: MFCD00236019
CAS Number: 98437-24-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR