4-Chloro-7-(trifluoromethyl)quinoline

Supplier: ACTIVATE SCIENTIFIC GMBH

ACTIAS2572G5EA 0 CHF
ACTIAS2572G5
4-Chloro-7-(trifluoromethyl)quinoline
4-Chloro-7-(trifluoromethyl)quinoline
Formula: C₁₀H₅ClF₃N
MDL Number: MFCD00006775
CAS Number: 346-55-4
EINECS: 206-471-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR