1,1,3,3-Tetrabromoacetone

Supplier: ACTIVATE SCIENTIFIC GMBH

Synonyms: 1,1,3,3-Tetrabromo-2-propanone

ACTIAS20523G25EA 534 CHF
ACTIAS20523G25 ACTIAS20523G5
1,1,3,3-Tetrabromoacetone
1,1,3,3-Tetrabromoacetone
Formula: C₃H₂Br₄O
MDL Number: MFCD00059470
CAS Number: 22612-89-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR