2,5-Dichloropyrazine

Supplier: ACTIVATE SCIENTIFIC GMBH

ACTIAS1213G5EA 0 CHF
ACTIAS1213G25 ACTIAS1213G5 ACTIAS1213G1
2,5-Dichloropyrazine
2,5-Dichloropyrazine
Formula: Câ‚„Hâ‚‚Clâ‚‚Nâ‚‚
MW: 148.9781 g/mol
MDL Number: MFCD04117886
CAS Number: 19745-07-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR