2-Amino-5-ethoxypyrazine

Supplier: ACTIVATE SCIENTIFIC GMBH

Synonyms: 5-Ethoxypyrazin-2-amine

ACTIAS30749G5EA 0 CHF
ACTIAS30749G5 ACTIAS30749G1
2-Amino-5-ethoxypyrazine
2-Amino-5-ethoxypyrazine
Formula: C₆H₉N₃O
MW: 139.1600 g/mol
MDL Number: MFCD09909690
CAS Number: 647843-58-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR