2-Amino-3-iodopyrazine

Supplier: ACTIVATE SCIENTIFIC GMBH

ACTIAS33382G1EA 0 CHF
ACTIAS33382G1
2-Amino-3-iodopyrazine
2-Amino-3-iodopyrazine
Formula: C₄H₄IN₃
CAS Number: 344329-41-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR