2-Bromopyrazine

Supplier: ACTIVATE SCIENTIFIC GMBH

ACTIAS30373G5EA 0 CHF
ACTIAS30373G5 ACTIAS30373G1
2-Bromopyrazine
2-Bromopyrazine
Formula: C₄H₃BrN₂
MW: 158.9800 g/mol
MDL Number: MFCD08275680
CAS Number: 56423-63-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR