2-Ethoxypyrazine

Supplier: ACTIVATE SCIENTIFIC GMBH

ACTIAS24411G25EA 120 CHF
ACTIAS24411G25 ACTIAS24411G100
2-Ethoxypyrazine
2-Ethoxypyrazine
Formula: C₆H₈N₂O
MW: 125.15 g/mol
MDL Number: MFCD00014609
CAS Number: 38028-67-0
EINECS: 253-750-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR