Phthalazine

Supplier: ACTIVATE SCIENTIFIC GMBH

Synonyms: Benzo[d]pyridazine

ACTIAS21344G25EA 0 CHF
ACTIAS21344G5 ACTIAS21344G25
Phthalazine
Phthalazine
Formula: C₈H₆N₂
MW: 130.15
MDL Number: MFCD00006908
CAS Number: 253-52-1
EINECS: 205-963-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR